Order Entry
Netherlands
ContactUsLinkComponent
Anti-ITGAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3.9]
Anti-ITGAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3.9]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ITGAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3.9]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-0116-T025
  • 50-0116-T100
  • Antigen symbol:
  • ITGA10
  • ITGA10
  • Antigen name:
  • Integrin Alpha 10
  • Integrin Alpha 10
  • Antigen synonyms:
  • integrin alpha-X|integrin aX|ITGAX|AI449405|CD11 antigen-like family member C|N418|Cr4|Cd11c
  • integrin alpha-X|integrin aX|ITGAX|AI449405|CD11 antigen-like family member C|N418|Cr4|Cd11c
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 3.9
  • 3.9
  • Reactivity:
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3687
  • 3687
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together