Anti-Diacylglycerol kinase iota (DGKI) Rabbit Polyclonal Antibody [clone: 342CT9.4.7]
ABGEAP8123B
: Abgent
- Antibody type:Primary
- Antigen name:Amyloid P Component, Serum
- Antigen symbol:Diacylglycerol kinase iota (DGKI)
- Clonality:Polyclonal
- Clone:342CT9.4.7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal (900–1000 aa)
- Cross adsorption:No
- Form:Ascites
- Gene ID:325
- Antigen synonyms:HEL-S-92n|Pentraxin 2|SAP|PTX2
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:MNKPLLWISVLTSLLEAFAHTDLSGKVFVFPRESVTDHVNLITPLEKPLQNFTLCFRAYSDLSRAYSLFSYNTQGRDNELLVYKERVGEYSLYIGRHKVTSKVIEKFPAPVHICVSWESSSGIAEFWINGTPLVKKGLRQGYFVEAQPKIVLGQEQDSYGGKFDRSQSFVGEIGDLYMWDSVLPPENILSAYQGTPLPANILDWQALNYEIRGYVIIKPLVWV
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Purified His-tagged APCS protein(Fragment) was used to produced this monoclonal antibody.
- Pk:400 µl
Type: Primary
Antigen: Diacylglycerol kinase iota (DGKI)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse