Anti-STX3 Rabbit Polyclonal Antibody
ABGEAP2818C
: Abgent
- Antibody type:Primary
- Antigen name:Syntaxin 3
- Antigen symbol:STX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Seq:03
- Epitope:middle
- Antigen synonyms:STX3A|Syntaxin-3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: STX3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human