Order Entry
Netherlands
ContactUsLinkComponent
Anti-SUMO4 (M55 Wild type) Rabbit Polyclonal Antibody
Anti-SUMO4 (M55 Wild type) Rabbit Polyclonal Antibody
Catalog # ABGEAP1264A
Supplier:  Abgent
undefined
Anti-SUMO4 (M55 Wild type) Rabbit Polyclonal Antibody
Catalog # ABGEAP1264A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Small ubiquitin-like modifier 4
  • Antigen symbol:
    SUMO4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    M55 Wild Type
  • Cross adsorption:
    No
  • Antigen synonyms:
    SMT3H4|Small ubiquitin-like protein 4|SMT3 suppressor of mif two 3 homolog 2|Small ubiquitin-like modifier 4|SUMO-4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SUMO4 (M55 Wild type)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human