Anti-ACTA1 Rabbit Polyclonal Antibody
AVIVARP60969_P050
- Antibody type:Primary
- Antigen name:Actin, alpha 1, skeletal muscle
- Antigen symbol:ACTA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:58
- Antigen synonyms:ASMA|CFTD|NEM2|NEM3|ACTA|NEM1|CFTDM|CFTD1|MPFD
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ACTA1 (actin, alpha 1, skeletal muscle)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human