Anti-TAAR5 Rabbit Polyclonal Antibody
Catalog # AVIVARP59626_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Trace amine associated receptor 5
- Antigen symbol:TAAR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9038
- Antigen synonyms:PNR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TAAR5 (trace amine associated receptor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat