Anti-CPNE7 Rabbit Polyclonal Antibody
AVIVARP55052_P050
- Antibody type:Primary
- Antigen name:copine VII
- Antigen symbol:CPNE7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:361433
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CPNE7 (copine VII)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: