Anti-DYNC1I2 Rabbit Polyclonal Antibody
Catalog # AVIVARP51954_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Dynein cytoplasmic 1, intermediate chain 2
- Antigen symbol:DYNC1I2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:1781
- Antigen synonyms:IC2|DNCI2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DYNC1I2 (dynein, cytoplasmic 1, intermediate chain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: