Anti-UNC93B1 Rabbit Polyclonal Antibody
AVIVARP49979_T100
- Antibody type:Primary
- Antigen name:unc-93 homolog B1 (C. elegans)
- Antigen symbol:UNC93B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:81622
- Antigen synonyms:Unc-93B1|UNC93B|IIAE1|UNC93
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: UNC93B1 (unc-93 homolog B1 (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: