Anti-PRPF19 Rabbit Polyclonal Antibody
Catalog # AVIVARP43158_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:pre-mRNA processing factor 19
- Antigen symbol:PRPF19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:27339
- Antigen synonyms:PRP19|hPSO4|NMP200|SNEV|UBOX4|PSO4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PRPF19 (pre-mRNA processing factor 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: