Bestelling invoeren
Netherlands
ContactUsLinkComponent
Test kit, water hardness, DUROVAL® B
Test kit, water hardness, DUROVAL® B
Catalog # HEYL400030
CAS Number:  
undefined
Test kit, water hardness, DUROVAL® B
Catalog # HEYL400030
CAS Number:  
Specifications
  • VPE:
    1 KIT
  • Omschrijving:
    Water hardness determination kit, with ~100 tests and measuring time of ~2 min, accuracy of 0,05 °dH
Specifications
About this item
Test kit for the determination of the water hardness in very soft or softened water samples via titration with a liquid titration agent and a pipette accurately calibrated in hardness units.
Delivery: Includes measuring tube, liquid indicator and pipette calibrated from 0 to 2 °dH.