Krimpkousen voor PETG mediaflessen, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
These heat-shrink bands can be used to ensure the integrity of bottle contents. Simply apply the recommended application torque for Thermo Scientific Nalgene® closures and slide the heat-shrink band over the closure and bottle neck. Heat the band to shrink and secure contents with a tamper-resistant seal.
- Gamma stable bands include a perforated "tear strip" feature for easy removal
Packaging: Bands are packed 1000 per case (two zipper bags of 500 each).
Specifications
- Catalog No:
- NALG312160-0384
- NALG312160-0200
- NALG312160-0530
- NALG312160-0240
- Environmentally Preferable:
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Milieu certificering:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- ISO 14001 (Certificate Registration No. EMS 623400)