Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium hexacyanoferrate (II)
Potassium hexacyanoferrate(II) solution 7 g/l
Potassium hexacyanoferrate(II) solution 7 g/l
Potassium hexacyanoferrate(II) solution 7 g/l
31138.368
:
VWR Collection
:
Potassium hexacyanoferrate(II) solution 7 g/l
31138.368
:
VWR Collection
:
Pk:
5 L
Seq:
5190
:
:
368.35 g/mol
:
13943-58-3
:
C₆FeK₄N₆
:
MFCD00167022