Deutrium furnace kit, GTA 120
Supplier: VARIAN
Certificates
About this item
GTA 120 deuterium graphite furnace AA operating supplies kit.
Delivery: Includes graphite electrodes, 2 pairs, spare graphite shroud, Omega platform tubes, 5×10/pk, 100 µl syringe for programmable sampler dispenser PSD-120, 5 capillary assemblies for PSD 120, 5×10 ml plastic (LDPE) beakers for PSD, 2 ml sample vials, 2×1000/pk
Frequently Bought Together