Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Transport Cooling Containers
Transport Cooling Containers
Marketsource
Supplier:   SARSTEDT
Transport Cooling Containers
Supplier:  SARSTEDT
Marketsource
Certificates
Transport Cooling Containers
Transport Cooling Containers
2 Options Available Below
Product Details & Documents
The cool transport containers is suitable for safe cool transportation of medical and biological specimens. It can accommodate two sample tubes of 10 ml volume or any tubes of 16,5 mm Ø and 107 mm length.
The mailing container is filled with a coolant and should be frozen in a horizontal position at −12 °C before mailing. The styrofoam casing protects against warmth and any damage during transport. The container is not suitable for samples that are not allowed to freeze.
Product Family Options
Product Information
  • Availability
    Price
  • SARS95.1123
    Complete with coolant, styrofoam casing and rubber band
    Complete with coolant, styrofoam casing and rubber band
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 84,71 €
    84,71 €
    1 * 6 ST
       84,71 €
    Specifications:
    Marketsource
    Marketsource
  • SARS95.1011
    Styrofoam casing with rubber band
    Styrofoam casing with rubber band
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 22,63 €
    22,63 €
    1 * 10 ST
       22,63 €
    Specifications:
    Marketsource
    Marketsource