Tubing, Bioprene
Supplier: WATSON MARLOW
Certificates
About this item
Bioprene offers exceptionally long life and resistance to a wide range of chemicals. It is a first choice for applications including metering tablet-coating materials, pH control and media feed in fermentation and bioreactor metering, as well as pharmaceutical handling, dispensing, metering, transfer and filtration.
- UV opaque
- Fully weldable
- No odour
- Resistant to oxidising agents
Bioprene peristaltic pump tubing is fully approved for food and pharmaceutical applications. Validated processes are supported. Like Marprene, It lasts at least 10 times as long as tubing in other materials. Long-life tubing means stoppages for a tube change are rare events, easily fitted in with general maintenance.
Temperature resistant from 5 to 80 °C
Hardness (Shore durometer A): 66
Frequently Bought Together