Clamps, three pronged
Catalog # RLAA1020/1
Supplier: R L ENTERPRISES
Specifications
- Spanning width:90 mm
- Description:With spring loaded jaws
- Seq:02
- Pk:1
Specifications
About this item
Nickel plated diecast aluminium alloy. Three prong clamp with nickel plated brass pummels and blue plastic headed nickel plated steel twin screw pivoting on a stainless steel pin.
- Jaws coated with chemical resistant plastic
- Spring loaded jaws (only RLAA1020/1)
- Clamp is designed to hold articles within the jaws
- For use with articles up to 90 mm diameter