Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Wipes
Sponges
Sponge
Sponge
Sponge
Catalog # KING060308
Supplier:
KING DISPOSABLES
Expand image
Sponge
Catalog # KING060308
Supplier:
KING DISPOSABLES
Specifications
Colour:
Yellow/green
Description:
Sponge, dual pad
Pk:
2
Specifications
About this item
Product Details
Synthetic sponge with green scouring pads.
Product Details