Self Seal Sterilisation Pouches, Transparent
Catalog # DIUKAU660-32
Supplier: DISTRIBUTOR
Specifications
- Description:Self-seal sterilization pouches, transparent
- L×W:130×254 mm
- Seq:02
- Pk:200
Specifications
About this item
Sterlising pouches suitable for autoclave or ethylene oxide gas sterilisation.
- The pouch has a two-ply polyester/polypropylene laminated transparent front bonded to a medical grade white sterilisation paper
- Dual arrows are printed on each pouch, pink changes to brown under normal autoclaving conditions and blue changes to yellow when gas sterilised
- A self-seal adhesive band is used to seal the pouch