Surface disinfectants, concentrates, Anistel
Leverancier: TRISTEL SOLUTIONS
Certificaten
Over dit artikel
Anistel is a surface disfectant used for high level animal health at the ratio of 1:100 for broad spectrum protection against Canine parvovirus, Feliine calivirus and a wide array of micro-organisms.
- Choice of fragrance
- 2 simple dilution levels 1:100 for high risk areas and 1:200 for general cleaning
- 5 minute contact time
Bactericidal, tuberculocidal, fungicidal, and virucidal.
Caution: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together