Strong cation exchange HPLC column, ProPac™ SCX-20
Supplier: Thermo Fisher Scientific
Certificates
About this item
This column is a strong cation exchange column designed specifically to provide high-resolution separation of proteins.
- pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity
- Designed for use with a standard-bore HPLC system having a gradient pump module, injection valve and a UV detector
- Compatible with typical mobile phases including MES, tris, phosphate or acetate buffers
Specifications
- Catalog No:
- 555-7499
- 555-7501
- Max. pressure:
- 3000 psi
- 3000 psi
- pH range:
- 2 - 12
- 2 - 12
- Temperature limit:
- 60 °C
- 60 °C
Specifications
Frequently Bought Together