Sartopore® 2 XLG capsules are self contained filter units that are especially designed for sterilising grade filtration in special applications of cell culture processes. Their unique heterogeneous double layer PES membrane combination is specifically developed to deal with the broad variety of contaminants in up- and downstream processing of biotechnology applications. They provide consistently high total throughput performance for biological fluid streams independent from media and process variations.
- Easy to use
- Flexible and cost saving
- Ideal for scale-up and scale-down
- Compatible with a pH range from 1 to 14 pH
PES membrane; support fleece, core and end caps: PP; O-rings: silicone; housing: PP; filling bell: PC
Sartopore® 2 XLG capsules are designed, developed and manufactured in accordance with a ISO 9001 certified Quality Management System. A Validation Guide is available for compliance with regulatory requirements.
- Catalog No:
- SARP5445307G8-SO-A
- SARP5445307G7-SO-A
- SARP5445307G0-SO-V
- SARP5441307G4-SS-B
- Max. pressure:
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 4 bar @ 20 °C; 2 bar @ 80 °C