Cation exchange HPLC columns, Dionex™, IonPac™ CS12A
Supplier: DIONEX
Certificates
About this item
CS12A columns are recommended for the fast isocratic separation of lithium, sodium, ammonium, potassium, magnesium and calcium from the sample matrices.
- Compatible with organic solvents
- High temperature
- High acid concentrations
Sample matrices are like environmental and drinking water, soil extracts, acid digest and power plant waters treated with ammonium or morpholine, and also industrial solvents.
Frequently Bought Together