Direct thermal Cryo-Tags® labels
Supplier: DIVERSIFIED BIOTECH
Certificates
About this item
These Cryo-Tags® polypropylene cryo labels accept printing from direct thermal printers such as Dymo® LabelWriter® to print popular barcode formats for fast easy sample identification.
- Temperature resistance from −196 to +70 °C
- Freezable in liquid and vapour phase nitrogen
- Adhere to most plastics, glass and metals without cracking, peeling or degrading, scuffing or smearing
Tubes labeled with direct thermal Cryo-Tags® easily slide in and out of centrifuge rotors without binding.
Caution: Only compatible with US model Dymo® labelWriters®.
Frequently Bought Together