Filter tips, Ovation®
These tips are made from 100% pure virgin PP. They contain a hydrophobic filter which acts as a barrier to aqueous liquids and aerosols, protecting the pipette and sample from trace amounts of carryover.
Frequently Bought Together