Order Entry
Netherlands
ContactUsLinkComponent
Sodium bis(2-methoxyethoxy)aluminium hydride 70% ∼3.6 mol/L in toluene
Flammable
Corrosive
Irritant
Health
Sodium bis(2-methoxyethoxy)aluminium hydride 70% ∼3.6 mol/L in toluene
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Sodium bis(2-methoxyethoxy)aluminium hydride 70% ∼3.6 mol/L in toluene
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (NBS Method):
    68.0 to 77.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together