
Iron(III) trifluoromethanesulphonate ≥90%, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63295-48-7
:
Formula:C₃F₉FeO₉S₃
MDL Number:MFCD15144785
:503.06 g/mol
Melting Pt:183 °C
Flash Pt:110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together