Fmoc-Pro-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:71989-31-6
:
Formula:C₂₀H₁₉NO₄
MDL Number:MFCD00037122
:337.38 g/mol
EINECS:276-259-0
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 99.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:116.0 to 121.0 deg-C
Specification Test Results
Frequently Bought Together