2,5-Dimethylpyrrole
Catalog # MOLE36723681-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:95.1424 g/mol
EINECS:210-913-8
CAS Number:625-84-3
Formula:C₆H₉N
MDL Number:MFCD00005223