
1-(Heptafluorobutyryl)imidazole ≥97.0% (by GC, titration analysis) acylating agent
Supplier: TCI
Certificates
About this item
CAS Number:32477-35-3
:
Formula:C₇H₃F₇N₂O
MDL Number:MFCD00014503
:264.10 g/mol
Flash Pt:77°C
EINECS:251-063-8
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid to slightly cloudy liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Effect test of GC derivalization:Effective as derivatization agent Dodecylamine
Specification Test Results
Frequently Bought Together