
6,6'-Dimethyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4411-80-7
:
Formula:C₁₂H₁₂N₂
MDL Number:MFCD00059776
Boiling Pt:286°C
:184.2371 g/mol
EINECS:224-566-5
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:88.0 to 95.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together