1,2-Diphenoxyethane ≥99.0% (by GC)
Catalog # TCIAD0866-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:95.0 to 98.0 deg-C
Specification Test Results
About this item
:
:214.26 g/mol
Boiling Pt:185°C
EINECS:203-224-9
CAS Number:104-66-5
Formula:C₁₄H₁₄O₂
MDL Number:MFCD00003039