
1,3-Dimethylimidazolium dimethylphosphate ≥97.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:654058-04-5
:
Formula:C₇H₁₅N₂O₄P
MDL Number:MFCD08275369
:222.18 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
Frequently Bought Together