2-Hexyloxybenzaldehyde 96%
Catalog # L08988.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:206.29 g/mol
Boiling Pt:132…134 °C (0.5 mmHg)
Storage Temperature:Ambient
CAS Number:7162-59-6
Formula:C₁₃H₁₈O₂
MDL Number:MFCD00014131