Didodecyl ether ≥95.0% (by GC)
Catalog # TCIAD0994-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 95.0 %
- Solubility in Toluene:very faint turbidity
Specification Test Results
About this item
:
Boiling Pt:190°C
EINECS:224-895-4
CAS Number:4542-57-8
Formula:C₂₄H₅₀O
MDL Number:MFCD00026590