
N,N'-Diethyl-1,6-diaminohexane ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13093-05-5
:
Formula:C₁₀H₂₄N₂
MDL Number:MFCD00026707
:172.3110 g/mol
Boiling Pt:228°C
EINECS:236-006-7
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together