
Benzhydrylamine ≥97.0% (by GC, titration analysis)
: TCI
:91-00-9
:
:C₁₃H₁₃N
:MFCD00008059
:183.25 g/mol
:14,01076
:304°C
:202-032-2
:GHS07
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Frequently Bought Together