DL-Phenylalaninol ≥98.0% (by GC, titration analysis)
TCIAP1791-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
- Specific rotation [a]20/D:-1.0 to +1.0 deg (C=5, 1mol/L HCl)
: 2-Amino-3-phenyl-1-propanol, DL-2-Amino-3-phenyl-1-propanol, 2-Amino-3-phenylpropan-1-ol
:
:151.21 g/mol
:3259
:GHS05
:16088-07-6
:MFCD00064298
:C₉H₁₃NO
:MFCD00066689
:130°C
:8 III