

N-Lauroylsarcosine sodium salt hydrate ≥98.0% (by total nitrogen basis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:137-16-6
:
Formula:H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MDL Number:MFCD00042728
Merck Index:14,04368
Melting Pt:46°C
Storage Temperature:Ambient
EINECS:205-281-5
UN:2811
ADR:6.1,II
:GHS05
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 % (calcd.on anh.substance)
- Water:max. 7.0 %
- Suitability for protein-analysis:to pass test
Specification Test Results
Frequently Bought Together