N(α)-Cbz-D-tryptophan ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2279-15-4
:
Formula:C₁₉H₁₈N₂O₄
MDL Number:MFCD00037945
:338.4 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:121.0 to 125.0 deg-C
- Specific rotation [a]20/D:+8.0 to +13.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together