Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-4-methylthiophene-3-carboxamide
2-Amino-4-methylthiophene-3-carboxamide
2-Amino-4-methylthiophene-3-carboxamide
2-Amino-4-methylthiophene-3-carboxamide
MAYBGK00199.10
:
Thermo Scientific
:
2-Amino-4-methylthiophene-3-carboxamide
MAYBGK00199.10
:
Thermo Scientific
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
156.20 g/mol
:
4651-97-2
:
C₆H₈N₂OS
:
MFCD00052594