Order Entry
Netherlands
ContactUsLinkComponent
1-(4-Nitrophenylazo)-2-naphthol
Irritant
1-(4-Nitrophenylazo)-2-naphthol
Catalog # TCIAP0586-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1-(4-Nitrophenylazo)-2-naphthol
Catalog # TCIAP0586-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Lambda max.:
    485.0 to 489.0 nm (Toluene)
  • Absorbance (E1%1cm):
    min. 800 (Toluene, 485.0 to 489.0 nm)

Specification Test Results

About this item

 :
 :293.28 g/mol
EINECS:229-093-8
 :GHS07
CAS Number:6410-10-2
Formula:C₁₆H₁₁N₃O₃
MDL Number:MFCD00003908