2,6-Diacetoxynaphthalene ≥99.0% (by GC)
Catalog # TCIAD1739-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:176.0 to 179.0 deg-C
Specification Test Results
About this item
:
CAS Number:22426-47-7
Formula:C₁₄H₁₂O₄
MDL Number:MFCD00035955