Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-Nitrobenzenesulphenyl chloride ≥95.0% (by GC, titration analysis) N-protecting agent, for peptide research
 :  TCI
 
undefined
 
2-Nitrobenzenesulphenyl chloride ≥95.0% (by GC, titration analysis) N-protecting agent, for peptide research
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Purity (Argentmetric Titration):
    min. 95.0 %
  • Melting point:
    74.0 to 77.0 deg-C

 

Frequently Bought Together