
2-Nitrobenzenesulphenyl chloride 98%
:7669-54-7
:
:C₆H₄ClNO₂S
:MFCD00007128
:70…75 °C
:Refrigerator
:231-644-2
:1759
:8,II
:189.62 g/mol
:GHS05
- Appearance:Yellow to golden Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:70 to 75 °C
- HPLC:≥97.5 %
Frequently Bought Together