Order Entry
Netherlands
ContactUsLinkComponent
3-Sulphobenzoic acid monosodium salt ≥95.0% (by titrimetric analysis)
Irritant
3-Sulphobenzoic acid monosodium salt ≥95.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
3-Sulphobenzoic acid monosodium salt ≥95.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 95.0 %
  • Solubility in Water:
    very faint turbidity

Specification Test Results

Frequently Bought Together