1,8-Diiodoanthracene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:189105-78-0
:
Formula:C₁₄H₈I₂
Storage Temperature:Freezer
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:199.0 to 203.0 deg-C
Specification Test Results
Frequently Bought Together