
1,2-Ethanedisulphonic acid dihydrate ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5982-56-9
:110-04-3
Formula:C₂H₆O₆S₂·2H₂O
MDL Number:MFCD26793415
:226.2260 g/mol
Merck Index:14,03724
EINECS:203-732-0
UN:2585
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (Neutralization titration):min. 95.0 % (calcd.on anh.substance)
- Melting point:111.0 to 115.0 deg-C
- Water:14.0 to 17.0 %
Specification Test Results
Frequently Bought Together