Order Entry
Netherlands
ContactUsLinkComponent
 
1,2-Bis(carboxymethylthio)ethane ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
1,2-Bis(carboxymethylthio)ethane ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    109.0 to 111.0 deg-C
  • Solubility in hot Water:
    almost transparency

 

Frequently Bought Together