2,3,5-Triphenyltetrazolium iodide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT0854-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
CAS Number:13014-90-9
Formula:C₁₉H₁₅IN₄
MDL Number:MFCD00060028